Learn More
C&G Containers 950CC AMB WM 53-400 WHT PTFE
SDP
Supplier:  C&G Containers LAP032237000
Encompass_Preferred
Log-ic Multi-Use USB Data Logger (26 uses) with wireless RFID technology to monitors temperature from -30degreesC to +75degreesC using s steel probe. Programmable. Tight accuracy of +/-0.5degreesC. Included software delivers professional reports.
Catalog No. 50156283