Learn More
Ace Glass, Inc. 9592 FLASK RB W/THREAD JOINT 9592 FLASK RB W/THREAD JOINT
SDP
Supplier:  Ace Glass, Inc. 959215
Encompass_Preferred
Microscale, single-neck, round-bottom reaction flask with outer joint externally threaded to allow an O-RING-CAP-SEAL connection with mating male joint. This type connection eliminates the need for clamps while offering the positive leak-tight seal of ground glass. Supplied with threaded cap with hole, one PTFE-faced silicone rubber septum and one O-Ring. 25mL 14/10 round bottom flask
Catalog No. NC9017828