Learn More
SEQUENCE BIOTECH LLC Precision Series 96 Channel Benchtop Pipettor - 1uL-20uL Working Volume.
For use with Sorenson OneTouch Pipette Tips.
Includes Pelican shipping case
Supplier: SEQUENCE BIOTECH LLC BMS5400020P
Formerly known as the Sorenson Benchtop Pipettor, Sequence Biotech now manages all production of the "Precision Series" Benchtop Pipettor. This model is optimized for use with Sorenson OneTouch Pipette Tips.
Key Applications:
ELISA Assays: The Precision Series is ideal for ELISA (Enzyme-Linked Immunosorbent Assays).
Cell Culture/Media Changes: Maintaining healthy cell cultures requires regular media changes, and this pipettor ensures efficient, precise transfers.
PCR Prep and Cleanup: For PCR amplification and subsequent cleanup, this device ensures accurate preparation and removes unwanted components for downstream applications like cloning or sequencing.
Plate-to-Plate Transfers: ensuring accurate and rapid liquid handling across 96-well plates. Ideal for high-throughput labs, it provides precise volume control and seamless operation.
Weighs only 20 lbs.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.