Learn More
SEQUENCE BIOTECH LLC LTS™-Plus 20uL w/ Pelican Case
(compatible with LTS™ Style Pipette Tips including Thermo Scientific™SoftFit-L™) - 1-20uL Working Volume.
Includes Pelican Case for Shipping and Service
Supplier:  SEQUENCE BIOTECH LLC LTSPLUS20P
Encompass_Preferred
Sequence Biotech introduces the LTS™-Plus 96/384 Channel Benchtop Pipettor. Designed for use with LTS™ or LTS™ compatible pipette tips. Working volume of 1-20uL.
Includes Pelican Case for Shipping and Service.
Key Applications:
ELISA Assays: The Precision Series is ideal for ELISA (Enzyme-Linked Immunosorbent Assays).
Cell Culture/Media Changes: Maintaining healthy cell cultures requires regular media changes, and this pipettor ensures efficient, precise transfers.
PCR Prep and Cleanup: For PCR amplification and subsequent cleanup, this device ensures accurate preparation and removes unwanted components for downstream applications like cloning or sequencing.
Plate-to-Plate Transfers: ensuring accurate and rapid liquid handling across 96-well plates. Ideal for high-throughput labs, it provides precise volume control and seamless operation.
Weighs only 20 lbs.
Catalog No. NC3311229
Only null left
Add to Cart
Request a Quote