Learn More
Thomas Scientific 96 Well 2mL Deep Well Plate, Round Bottom, Sterile, 50/cs
SDP
Supplier:  Thomas Scientific 1149J76
Encompass
Ideal for sample collection, preparation, and long-term storage. HTS-friendly-compatible with most liquid-handling robots. Centrifugation to 4,000 x G.
Catalog No. NC1833740