Learn More
Analytical Sales & Services EquaVAP 96-Well Blowdown Evaporator, Even Evaporation, High Reproducibility, 4.558288086873203 Qty
Supplier:  Analytical Sales & Services 23096
Encompass
The Analytical Sales & Services EquaVAP 96-Well Blowdown Evaporator is a high-throughput sample concentration system designed for efficient and uniform evaporation across 96-well plates. With features like even gas distribution, high reproducibility, and robust construction, it ensures reliable results while its compact design fits standard fume hoods. Ideal for scientific labs, it requires only a nitrogen supply and power connection, making it easy to operate and versatile with various well plate types.
Catalog No. NC1425008