Learn More
Thomas Scientific 96 Deep Well HTS Reservoir, White PS, Sterile (1/pack, 50/case + 2 Reservoir Holders)
SDP
Supplier:  Thomas Scientific 1171H96/DEL
Encompass_Preferred
8 Channel, 96 or 384 well format Shallow well: 75mL Deep well: 275mL Available in Natural and White Virgin Polystyrene Reusable SBS Footprint Holder
Catalog No. NC0272615