Learn More
Thomas Scientific 96 Well ELISA Plate, Undetachable, High Binding, Clear
SDP
Supplier:  Thomas Scientific 1193A15
Encompass_Preferred
Designed for ELISA method Medical grade polystyrene Conform to SBS/ ANSI Both detachable and undetachable plates are available Flat and transparent well bottom
Catalog No. 50-145-8305