Learn More
Analytical Sales & Services 96-Well Lightweight PP Well Tray, Inert, Excellent Chemical Resistance, Compatible with Automation
Supplier:  Analytical Sales & Services 96342
Encompass
The 96-Well Lightweight Polypropylene Well Tray is designed for high-throughput analytical applications. With 96 wells, pre-assembled glass inserts, and exceptional thermal stability, it minimizes sample adsorption while providing chemical resistance. Its lightweight design and automation compatibility make it ideal for various laboratory processes, enhancing efficiency in sample preparation and chromatography.
Catalog No. NC1063261