Learn More
Electron Microscopy Sciences NUNC™ 96 Well Optical Bottom Plate Black, Cell Culture Sterile
Supplier: Electron Microscopy Sciences 6481005
The NUNC Coverglass Optical Bottom 96 Well Plates have a polymer bottom and combine the optical clarity of virgin crystalline polystyrene with optimal surface for a wide range of HTS applications. The optical clarity allows for easy viewing of well contents. Footprint compatible with standard equipment and automated systems. It has a flat bottom well geometry for plate reader access and working ranges from 50-200 uL/well. White upper structure with Polymer bottom.
Color: Black
Surface: Cell Culture
Sterile: Sterile