Learn More
Lonza Walkersville 96-well plates intended for use with LAL-based endotoxin assays.The plates are certified to contain <0.0005 EU/well endotoxin and are packaged 5 plates/pack, 10 packs/case.
Supplier:  Lonza Walkersville 00313405
Encompass
These 96-well plates can be used with the Kinetic-QCL Kinetic Chromogenic LAL Assay and the PYROGENT 5000 Kinetic Turbidimetric LAL Assay. The plates are certified to contain <0.0005 EU/well endotoxin and are packaged 5 plates/pack, 10 packs/case.
Catalog No. NC3827344