Learn More
LUNA NANOTECH INC 96-Well Silica Membrane Plate, Semi-Skirted
Supplier: LUNA NANOTECH INC USP096S
Speed up your RNA and DNA processing purification by going high throughput!
Each well features a thick silica membrane to maximize nucleic acid binding and recovery of RNA and DNA from many sample types.
Binding capacity:
- RNA: 200 µg
- Plasmid DNA: up to 20 µg
- PCR Product: up to 20 µg
- Genomic DNA: 100 µg
- Our Purification Plate allows the user to process up to 96 samples simultaneously.
- Processing can be done via centrifugation or vacuum manifold.
- Available in Full-skirted and Semi-skirted format.