Learn More
Bellco Glass, Inc. 96 Well Square well Silicone Sealing Mat, non-sterile, 10/pk, 50/cs
SDP
Supplier:  Bellco Glass, Inc. 451400603
Encompass_Preferred
96 Well Square well Silicone Sealing Mat, non-sterile, 10/pk, 50/cs. sealing mats minimize evaporation in a wide range of temperatures in storage applications. Made of research-grade silicone, they can be autoclaved, and following a bleach wash and ethanol rinse protocol, they can also be reused. • Easy Press and Mini sealer offer easier-operating and time-consuming options.• Good airtightness offer unsurpassed protection for volatile solvents.• Chemically resistant, pierceable thermoplastic elastomer well caps are excellent for strong to -80℃.• Silicone pre-slit well caps permit a pipette tips or probe to enter the well without damage to the cap.
Catalog No. 50-233-0749