Learn More
Alkali Scientific 96 Well White Clear Bottom Cel
SDP
Supplier:  Alkali Scientific TPW4096
Encompass
Our 96-well cell culture plates are versatile and ideal for a variety of lab applications including cloning experiments sample incubations and tissue culture-based analytical systems These plates along with plate sealers ensure flexibility reliability and reproducibility throughout the cell culture process Available in a range of surfaces and formats they are designed to meet the diverse requirements of cell attachment and growth Our cell culture plates promote optimal cell health and consistent results Some tissue culture plates feature excellent stackability and radiation sterilization for smooth handling Glass-bottom well plates allow for easy live cell imaging without transfer ensuring accurate results with superior flatness Key features Suitable for fluorescence and luminescence assays Designed to prevent cross-contamination Effective gas exchange lid design Alphanumeric labeling for easy identification Promotes homogeneous cell culture Flat-bottom wells USP Class VI complia
Catalog No. 50-302-8929