Learn More
Thomas Scientific 96W PLT PS ST TC WHT LID 32/CS
SDP
Supplier:  Thomas Scientific 1184J24
Encompass_Preferred
96 Well GreinerMicroplate, White, F-Bottom, Chimney Well, TC Treated, with Lid, Sterile; Growth area, 34mm(2); Total volume, 392ul; Working volume, 25-340ul.
Catalog No. 50-320-854