Learn More
Lonza Walkersville Cell Line Optimization 96-well Nucleofector Kit
Supplier:  Lonza Walkersville V4SC9096
Encompass
The Cell Line Optimization 96-well Nucleofector Kit is the ideal tool to conveniently and rapidly determine Nucleofection conditions for cell lines lacking an Optimized Protocol for the 4D-Nucleofector 96-well Unit.Benefits:Convenient and rapid determination of optimal Nucleofection conditions for a broad range of primary cells within one experimentOptimal Nucleofection conditions determined on the 4D-Nucleofector 96-well Unit are transferable to the 4D-Nucleofector System or the 384-well HT Nucleofector SystemApplications:Determination of Nucleofection conditions for cell lines lacking an Optimized Protocol for the 4D-Nucleofector 96-well UnitContent:0.675 mL Nucleofector Solution SE0.675 mL Nucleofector Solution SF0.675 mL Nucleofector Solution SG3 x 0.15 mL Supplement 150 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0)1 x 96-well Nucleocuvette Plate
Catalog No. NC0111672