Learn More
Medchemexpress LLC 98N12-5 | 917572-74-8 | 98.0% | 1343.22 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY112772A100MG
Encompass_Preferred
98N12-5 is a novel multi-tail ionizable lipid. It has been utilized for efficient in vivo siRNA-delivery to the liver.
  • Novel multi-tail ionizable lipid.
  • Used for efficient in vivo siRNA-delivery.
  • Specifically targets the liver for siRNA delivery.
Catalog No. 50-004-37179