Learn More
Medchemexpress LLC 9H-Carbazole-3-carba 1g
SDP
Supplier:  Medchemexpress LLC HYW1068601G
Encompass_Preferred
9H-Carbazole-3-carbaldehyde is an anticancer agent that can be isolated from the stems of Lansium parasiticum (Lour ) Skeels 9H-Carbazole-3-carbaldehyde shows significant cytotoxic activity in a variety of tumor cell lines especially against H1299 and SMMC-7721 lung cancer cells with IC50 values of 6 19-26 84 g/mL[1]
Catalog No. 50-002-05081