Learn More
Medchemexpress LLC 9H-Carbazole-9-ethanol, 3,6-dibromo-a-[[(3-methoxyphenyl)amino]methyl]- | 313268-18-7 | 98.3% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12885650MG
Encompass_Preferred
P7C3-OMe is a pro-neurogenic compound that offers therapeutic benefits for neuropsychiatric and neurodegenerative diseases. The R-enantiomer of this compound demonstrates significantly greater activity compared to the S-enantiomer.
  • Pro-neurogenic compound
  • Therapeutic benefits in neuropsychiatric and neurodegenerative diseases
  • R-enantiomer is significantly more active
Catalog No. 50-003-38964