Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9H-Fluoren-9-one, 2-fluoro- | 343-01-1 | 98.0% | 198.20 | 250 MG
SDP

Supplier:  Medchemexpress LLC 250MGHYW009829

Encompass_Preferred

2-Fluorofluoren-9-one is a chemical compound with the molecular formula C13H7FO and a molecular weight of 198.20. It is presented as a light yellow to yellow solid and is suitable for various research applications.

  • Has a purity of 98.03% as determined by LCMS
  • Consistent with structure by ¹H NMR spectrum and LCMS
  • Recommended storage for powder: -20°C for 3 years or 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months or -20°C for 1 month
  • Identified by CAS number 343-01-1

Catalog No. 50-004-51294


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.