Learn More
Medchemexpress LLC (((9H-Fluoren-9-yl)methoxy)carbonyl)-D-histidine | 157355-79-8 | MFCD00237022 | 377.39 g·mol⁻¹ | C21H19N3O4 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW048701
Encompass_Preferred
(((9H-Fluoren-9-yl)methoxy)carbonyl)-D-histidine is an Fmoc-protected D-enantiomer of the amino acid histidine used as a building block in peptide synthesis and related peptide chemistry. It is supplied in small research quantities for incorporation into synthetic peptides and peptide-coupling reactions.
  • Fmoc-protected D-histidine for stereospecific incorporation
  • Suitable for solid-phase peptide synthesis workflows
  • Useful for peptide coupling and residue-specific modifications
  • Available in small-scale research sizes (example: 250 mg)
  • Molecular weight 377.39 g·mol⁻¹; molecular formula C21H19N3O4
Catalog No. 50-002-08467