Learn More
Medchemexpress LLC 9H-purin-6-amine, 9-(2-deoxy-2-fluoro-β-D-arabinofuranosyl) | 20227-41-2 | MFCD12911815 | 99.9% | 269.24 | C10H12FN5O3 | 100 MG

Supplier: Medchemexpress LLC HYW251687100MG
2'-Deoxy-2'-fluoroarabinoadenosine is a fluorinated deoxyadenosine nucleoside analogue used in research on antiviral and anticancer mechanisms and as a building block for modified oligonucleotides.
- High purity: 99.92% by LCMS.
- White to off-white solid appearance.
- Molecular weight 269.24; formula C10H12FN5O3.
- CAS number 20227-41-2 for clear identification.
- Storage: keep at 4°C protected from light; in solvent store at -80°C (6 months) or -20°C (1 month).
- Analytical data consistent with structure (1H NMR, LCMS).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.