Learn More
Diversified Biotech Inc 9L ICEPAN,BLU,5/CS
Supplier:  Diversified Biotech Inc IPAN3000CS
Encompass
IcePan, Cooling, Triple Density Expanded Polystyrene Ice lasts 30% longer, sweatless, durable, tolerates dry ice, liquid nitrogen, slurry 9L GLACIERBRAND ICE PAN, BLUE
Catalog No. NC1605898