Learn More
SPT LABTECH INC Spool Of 13500 Non-Sterile Pipettes 9mm Pitch HV
Supplier:  SPT LABTECH INC 415003025
Encompass
Disposable positive-displacement pipettes give superior accuracy for a wide range of liquid viscosities. Volume range of 500 nL to 5μL, suitable for addressing 96-, 384- and 1,536-well plates.
Catalog No. NC1768402