Learn More
Leica Biosystems Leica Nozzle 21G - Dispenser nozzle for CV5030 coverslipper
Supplier:  Leica Biosystems 14047840157
Encompass_Preferred
The Leica Biosystems Nozzle 21G (MPN 14047840157) is a replacement dispenser nozzle designed for use with the Leica CV5030 glass coverslipper. It delivers precise, low-volume dispensing for consistent cover glass application during automated coverslipping operations.
  • Provides fine dispensing control for coverslipping.
  • 21-gauge size for precise, low-volume delivery.
  • Designed for use with automated glass coverslippers.
  • Sold individually for straightforward replacement.
  • Helps maintain consistent cover glass placement and adhesion.
Catalog No. NC0356127