Learn More
Leica Biosystems Leica Biosystems power cord - NEMA 5-15 (US) to IEC C13 appliance connector (replacement power cord)
Supplier:  Leica Biosystems 14041157009
Encompass_Preferred
Replacement power cord for compatible laboratory instruments. Connects equipment with an IEC C13 inlet to North American mains via a NEMA 5-15 plug. Intended as a general-purpose appliance lead for instruments requiring an IEC C13 connection.
  • Plug type: NEMA 5-15 (male)
  • Connector: IEC 60320 C13 (female)
  • Suitable for equipment with an IEC C13 inlet
  • Grounded three-conductor cable for safety
  • Available in regional plug variants; check local compatibility and certifications
Catalog No. NC1458128