Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter Beckman Coulter CD158e1/e2-PE antibody, Clone Z27.3.7, 1 mL (IM3292)

Supplier:  Beckman Coulter IM3292

Encompass

CD158e1/e2-PE is a PE-conjugated monoclonal antibody (clone Z27.3.7) for detection of CD158e1 and CD158e2 (KIR3DL1/KIR3DS1) on human natural killer (NK) cells and subsets of CD8+ T cells by flow cytometry. Supplied in liquid format as an analyte specific reagent, 1 mL.

  • Target species: Human.
  • Specificity: CD158e1 and CD158e2.
  • Clone: Z27.3.7.
  • Fluorochrome: PE (phycoerythrin).
  • Isotype: IgG1, mouse.
  • Format: Liquid, 1 mL.
  • Regulatory status: ASR (analyte specific reagent).
  • Recommended use: Flow cytometry.

Catalog No. NC2683284


May include imposed supplier surcharges.
Add to Cart
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.