Learn More
ATCC ATCC PDM-33 (HCM-CSHL-0082-C25) - Human primary pancreatic ductal adenocarcinoma patient-derived tumor model
Supplier:  ATCC HCMCSHL0082C25
Encompass
A patient-derived primary tumor model established from human pancreatic ductal adenocarcinoma tissue for use in cancer research and translational studies.
  • Derived from primary pancreatic ductal adenocarcinoma tissue.
  • Human (Homo sapiens) origin.
  • Suitable for cancer research, immunotherapy, and cell and gene therapy studies.
  • Biosafety level 1.
  • Supplied as an individual model (packaging: each).
  • Patient-derived model representing tumor heterogeneity.
Catalog No. NC3660184