Learn More
Bachem Amyloid Beta Protein (1-42)
SDP
Supplier:  Bachem 4014447.0010
Encompass_Preferred
Amyloid Beta Protein (1-42), H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala-OH, 107761-42-2, C203H311N55O60S, Mr 4514.10, 10mg. Ab42, b-Amyloid (1-42). Ab 1-42, 42-residue fragment of amyloid precursor protein, has been found to be a major constituent of the senile plaques formed in the brains of patients with Alzheimer's disease and late Down's syndrome. Ab 1-42 readily forms neurotoxic oligomers at physiological pH. On the other hand, the peptide shows antimicrobial activity. The sequence of this peptide corresponds to the sequence of human, bovine, canine, feline, ovine, guinea pig, and rabbit Ab42. The peptide has been used to detect amyloid b-protein multimers in the cerebrospinal fluid of Alzheimer's disease patients through fluorescence correlation spectroscopy.
Catalog No. 50-194-6576