Learn More
Medchemexpress LLC Aβ1-42 aggregation inhibitor 1 | 2439107-75-0 | 468.66 | 10 MG
SDP
Supplier:  Medchemexpress LLC 10MGHY150003
Encompass_Preferred
Aβ1-42 aggregation inhibitor 1 inhibits AChE (acetylcholinesterase) and BuChE (butyrylcholinesterase) with IC50 values of 2.64 μM and 1.29 μM, respectively. This inhibitor reduces self-mediated Aβ1-42 aggregation by 51.29% at a concentration of 25 μM, indicating its potential for anti-Alzheimer's disease research. Additionally, it can selectively form ligand-Cu2+ complexes with Cu2+ at 25 μM within 30 minutes.
  • Inhibits AChE (acetylcholinesterase) and BuChE (butyrylcholinesterase)
  • Reduces self-mediated Aβ1-42 aggregation
  • Potential for research into anti-Alzheimer's disease
  • Selectively forms ligand-Cu2+ complexes with Cu2+
Catalog No. 50-003-65612