Learn More
Medchemexpress LLC A 1120 50mg | 1152782-19-8 | 50MG
SDP
Supplier:  Medchemexpress LLC HY10763350MG
Encompass_Preferred
A 1120 is a high-affinity nonretinoid retinol-binding protein 4 (RBP4) antagonist with a Ki value of 8 3 nM A 1120 disrupts the interaction between RBP4 and its binding partner transthyretin[1 [2
Catalog No. 50-002-40293