Learn More
Medchemexpress LLC HY-19725 10mg , A-1155463 CAS:1235034-55-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1972510MG
Encompass_Preferred
HY-19725 10mg A-1155463 CAS: 1235034-55-5 A-1155463 is a highly potent and selective BCL-XL inhibitor with an EC50 of 70 nM in Molt-4 cell. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9386