Learn More
Selleck Chemical LLC A-1331852
SDP
Supplier:  Selleck Chemical LLC S780125MG
Encompass_Preferred
A-1331852 is a potent and selectiveBCL-XL inhibitor with Ki value less than 0 01 nM for BCL-XL and 6 nM 4 nM 142 nM for Bcl-2 Bcl-W MCL-1 respectively It may be useful in the treatment of cancer immune and autoimmune diseases
Catalog No. 50-000-04497