Learn More
Selleck Chemical LLC A-1331852 5mg 1430844-80-6
SDP
Supplier:  Selleck Chemical LLC S78015MG
Encompass_Preferred
A-1331852 is a potent and selectiveBCL-XL inhibitor with Ki value less than 0.01 nM for BCL-XL and 6 nM, 4 nM, 142 nM for Bcl-2, Bcl-W, MCL-1 respectively. It may be useful in the treatment of cancer, immune and autoimmune diseases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5269