Learn More
Selleck Chemical LLC A-366 10mg 1527503-11-2
SDP
Supplier:  Selleck Chemical LLC S757210MG
Encompass_Preferred
A-366 is a potent and selective G9a/GLP histone lysine methyltransferase inhibitor with IC50 of 3.3nM, exhibiting >1000-fold selectivity for G9a/GLP over 21 other methyltransferases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4959