Learn More
Medchemexpress LLC HY-16014 5mg Medchemexpress, A-385358 CAS:406228-55-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY160145MG
Encompass_Preferred
Medchemexpress, HY-16014 5mg A-385358 CAS:406228-55-5 A-385358 is a selective inhibitor of Bcl-XL with Kis of 0.80 and 67 nM for Bcl-XL and Bcl-2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1862