Learn More
Medchemexpress LLC A 410099.1, amine-Boc hydrochloride | 2374122-37-7 | 99.7% | 620.22 g/mol | C32H50ClN5O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1362695MG
Encompass_Preferred
A 410099.1, amine-Boc hydrochloride is a Boc-protected amine derivative of an IAP ligand supplied as the hydrochloride salt. It is intended as a functionalized building block for the synthesis of proteolysis-targeting chimeras (PROTACs), including BTK-targeting PROTACs, and is provided for research and medicinal chemistry applications.
  • High purity (99.7%) suitable for synthesis workflows.
  • Boc-protected amine functionality for facile deprotection and conjugation.
  • Hydrochloride salt form improves handling and stability.
  • Compatible with PROTAC assembly and medicinal chemistry derivatization.
  • Available in small mg packaging for research-scale use.
Catalog No. 50-002-22202