Learn More
Medchemexpress LLC Aβ42-IN-2 | 1914989-80-2 | 98.0% | 430.50 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13686610MM/1ML
Encompass_Preferred
Aβ42-IN-2 is a γ-secretase modulator with an IC50 of 6.5 nM for Aβ42, making it suitable for Alzheimer's disease research. This product is for research use only.
  • γ-Secretase modulator
  • IC50 of 6.5 nM for Aβ42
  • Used in Alzheimer's disease research
Catalog No. 50-003-48682