Learn More
Medchemexpress LLC HY-15488A 10mg Medchemexpress, A 438079 (hydrochloride) CAS:899431-18-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15488A10MG
Encompass_Preferred
Medchemexpress, HY-15488A 10mg A 438079 (hydrochloride) CAS:899431-18-6 A 438079 (hydrochloride) is a potent, and selective  P2X 7 receptor  antagonist with pIC 50  of 6.9. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2744