Learn More
Medchemexpress LLC A-485 Solution 10 mM * 1 mL in DMSO | CAS No. : 1889279-16-6
SDP
Supplier:  Medchemexpress LLC HY10745510MMSOL
Encompass_Preferred
A-485 is a potent and selective catalytic inhibitor of p300/CBP with IC50s of 9.8 nM and 2.6 nM for p300 and CBP histone acetyltransferase (HAT), respectively.
Catalog No. NC3354200