Learn More
Apexbio Technology LLC A-674563, 5mg. Cas: 552325-73-2 MFCD: MFCD11840461. Akt1/PKA/CDK2 inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC A86165
Encompass_Preferred
A-674563 is a potent and selective Akt1 inhibitor with IC50 value of 14 nM. A-67453 also inhibited PKA and Cdk2 with IC50 value of 16 nM and 46 nM. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5164