Learn More
Selleck Chemical LLC A-674563 HCl S2670-100mg
SDP
Supplier:  Selleck Chemical LLC S2670100MG
Encompass_Preferred
A-674563 HCl is an Akt1 inhibitor with Ki of 11 nM in cell-free assays modest potent to PKA and 30-fold selective for Akt1 over PKC
Catalog No. 50-311-2573