Learn More
Medchemexpress LLC A-7 hydrochloride (Ophobolin A) | 79127-24-5 | MFCD00058400 | 99.1% | 433.44 g/mol | C20H30Cl2N2O2S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1009145MG
Encompass_Preferred
A-7 hydrochloride (Ophobolin A) is the hydrochloride salt of a fungal sesterterpenoid that functions as a calmodulin antagonist and is used as a biochemical tool in cancer and calmodulin-related research.
  • Acts as a calmodulin antagonist in biochemical assays.
  • Useful for cancer-related cellular and molecular studies.
  • Provided as a solid, light yellow to yellow substance.
  • High reported purity (99.1%).
  • Supplied in small milligram quantities suitable for lab-scale experiments.
  • For research use only; not intended for clinical or human use.
Catalog No. 50-002-11586