Learn More
Medchemexpress LLC A 779 | 159432-28-7 | 99.3% | 872.97 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYP021650MG
Encompass_Preferred
A 779 is a specific antagonist of the G-protein coupled receptor (Mas receptor), which is an Ang1-7 receptor distinct from the classical AngII. It is intended for research use only.
  • Specific antagonist of the G-protein coupled Mas receptor
  • Inhibits the effect of Ang-(1-7)
  • Suppresses PCNA protein expression upregulated by Ang II
  • Retards Ang II-induced inflammatory responses in VSMCs
  • Blocks Ang-(1-7) effect on Akt and ERK1/2 phosphorylation
  • Elevates several serum bone markers (BALP, CTX, TRAcP 5b, OC, DPD)
  • Abolishes Ang(1-7) favorable effects on bone health
  • Eradicates captopril's protective effects on bone metabolism
  • Restores OVX effects on RANKL expression and the ACE-1/AngII/AT1R cascade
  • Down-regulates OPG expression and the ACE-2/Ang1-7/Mas pathway
Catalog No. 50-004-25898