Learn More
Selleck Chemical LLC A-803467 S2785-50mg
SDP
Supplier:  Selleck Chemical LLC S278550MG
Encompass_Preferred
A-803467 is a selective NaV1 8 channel blocker with IC50 of 8 nM blocks tetrodotoxin-resistant currents exhibits 100-fold selectivity against human NaV1 2 NaV1 3 NaV1 5 and NaV1 7
Catalog No. 50-311-1863