Learn More
Selleck Chemical LLC A-804598 10mg 1125758-85-1
SDP
Supplier:  Selleck Chemical LLC S872510MG
Encompass_Preferred
A-804598 is a selective P2X7R antagonist with high affinity at rat (IC50 = 10 nM), mouse (IC50 = 9 nM) and human (IC50 = 11 nM) P2X7 receptors. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8283