Learn More
Selleck Chemical LLC A-83-01
SDP
Supplier:  Selleck Chemical LLC S76921G
Encompass_Preferred
A-83-01 is a potent inhibitor of TGF- type I receptor (ALK5-TD) with IC50 of 12 nM A-83-01 also inhibits the transcription induced by activin/nodal type I receptor (ALK4-TD) and nodal type I receptor (ALK7-TD) with IC50 of 45 nM and 7 5 nM respectively Solutions are unstable and should be fresh-prepared
Catalog No. 50-000-00348