Learn More
Cayman Chemical A-834735 Degradnt 10mg
SDP
Supplier:  Cayman Chemical 1416510 MG
Encompass_Preferred
A common impurity observed during GC-MS analysis of samples containing A-834735, featuring an opened ring presumed to be produced during heating; gives rise to a prominent fragment ion that is 15 amu greater than the base peak of A-834735
Catalog No. 50-295-3604