Learn More
Medchemexpress LLC A 922500 | 959122-11-3 | MFCD12828780 | 99.3% | 428.48 | C26H24N2O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1003810MG
Encompass_Preferred
A 922500 is a potent, selective small-molecule inhibitor of diacylglycerol acyltransferase 1 (DGAT-1) supplied for research use. It is orally bioavailable and commonly used as a biochemical probe to study lipid metabolism and DGAT-1-related pathways.
  • Potent and selective DGAT-1 inhibition.
  • Orally bioavailable small molecule suitable for in vivo studies.
  • Provided with characterization data and an SDS for safe handling.
  • Available in multiple sizes and solution formats for experimental flexibility.
  • High purity to support reproducible biochemical assays.
Catalog No. 50-202-8616