Learn More
Medchemexpress LLC A 922500 | 959122-11-3 | MFCD12828780 | 99.3% | 428.48 | C26H24N2O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1003810MM 1ML
Encompass_Preferred
A 922500 (DGAT-1 Inhibitor 4a) is a potent, selective, orally bioavailable inhibitor of diacylglycerol acyltransferase 1 (DGAT-1) with low-nanomolar activity against human and mouse targets. It is supplied at high purity and offered both as solid material in multiple sizes and as a ready-to-use 10 mM solution in DMSO for in vitro work or formulation for in vivo studies.
  • Potent DGAT-1 inhibition (IC50 ≈ 9 nM human, 22 nM mouse).
  • High purity (>99%).
  • Available as a 10 mM solution in DMSO for direct use.
  • Soluble in DMSO at ≥50 mg/mL; in vivo formulation options documented.
  • Multiple solid package sizes for dosing flexibility.
Catalog No. 50-002-14589